Orders LinkContactUsLinkComponent
Cyclosporine, Reference Standards
Cyclosporine, Reference Standards
Cyclosporine Resolution Mixture, USP reference standard, Vial, 25 mg
Codice catalogo 1158650.
Nome del fornitore:   USP
Cyclosporine, Reference Standards
Codice catalogo 1158650.
Nome del fornitore:  USP
Numero CAS:  
Cyclosporine, Reference Standards
Cyclosporine Resolution Mixture, USP reference standard, Vial, 25 mg
Numero CAS:  
Price
1 430.00 SFr.
Log in per disponibilità
1 * 25 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Cyclosporine Resolution Mixture: This material is an approximate 100:1 (w/w) mixture of cyclosporine and cyclosporine U
Certifications: US Pharmacopea Reference Standard
Specifiche
Specifiche
  • Tipo di conf.:
    Vial
  • Conf:
    25 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Cyclosporine Resolution Mixture