Fluoresceina
100 µG
CHMCMAB045X
: Chemicon
: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- :2321-07-5
- :C₂₀H₁₂O₅
- :MFCD00005050
- :332.31 g/mol
- :13,04185
- :314…316 °C
- :Ambiente
- :219-031-8
- Conf:100 µG