Orders LinkContactUsLinkComponent
Cloning cylinders
Cloning cylinders
10 mm, 10 mm
Codice catalogo CHMCTR-1005
Nome del fornitore:   Chemicon
Cloning cylinders
Codice catalogo CHMCTR-1005
Nome del fornitore:  Chemicon
Cloning cylinders
10 mm, 10 mm
Price
SFr. 217.00
Log in per disponibilità
1 * 10 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Sterile
  • Ready to use
  • Can be stored at room temperature
Individual colonies of transfected cells can be isolated and picked from a plate containing many clones. These sterile cloning cylinders are greased on one end to allow the cylinder to seal to the plate. Cells are then dissociated within the cylinder, resuspended, and transferred to a new vessel as a homogeneous population of cells derived from a single colony.
Specifiche
  • Ø est:
    10 mm
  • Altezza:
    10 mm
  • Seq:
    02