Orders LinkContactUsLinkComponent
Termociclatori, Mastercycler® ep
Termociclatori, Mastercycler® ep
Mastercycler® ep cycleManager, incl. installation instructions,
comprehensive online-help, connection cable
Codice catalogo EPPE5349810.001
Nome del fornitore:   EPPENDORF
Termociclatori, Mastercycler® ep
Codice catalogo EPPE5349810.001
Nome del fornitore:  EPPENDORF
Termociclatori, Mastercycler® ep
Termociclatori, Mastercycler® ep
Mastercycler® ep cycleManager, incl. installation instructions,
comprehensive online-help, connection cable
Price
SFr. 4’530.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The Mastercycler® ep family of thermal cyclers are distinguished by having thermoblocks with extremely fast heat-ing and cooling speeds, and unique heated lid technology (ESP - Electronic Sample Protection), which avoids the preheating of samples and minimises unspecific annealing. The gradient option of each Mastercycler® ep system features SteadySlope® technology to ensure that the heating and cooling ramp rates are identical in both gradient and normal operation. Programming with the control panel is easy and intuitive, allowing novice users to get started quickly and safely.
  • Control panel or PC software driven
  • ESP heated lid
  • Available with manual or motorised lid
  • Space-saving footprint with sliding lid and no side air vents necessary
  • SteadySlope® technology ensures identical temperature control characteristics in both optimisation and rou-tine experiments, as well as a reliable transfer of the optimisation results to the routine application
For integration into fully automated environments, Mastercycler® ep models are also available with an optional motorised lid. For additional speed, the Mastercycler® ep gradient S incorporates a device driven Hot Start. The Mastercycler® ep models can be operated in various configurations and adjusted to the requirements and size of the laboratory - as a universal, stand alone device with control panel operation; as a ‘mini-satellite’ system consisting of a control panel and up to 5 Mastercycler ep models; as a PC supported, GLP conforming ‘PCR machine’; as a PC controlled Mastercycler ep network; or as high throughput units with motorised lids for automation integration.
Temperature control range of block: 4 to 99 °C
Interfaces: Centronics®, RS232, control panel, CAN_in/CAN_out
* Per il funzionamento sono necessari il pannello di controllo o il software epCycleManager; per il collegamento dei vari moduli termici in un'unica rete sono necessari i cavi di collegamento CAN-bus.
Accessories information: L'adattatore in situ può essere utilizzato per trasformare il Mastercycler® o il Mastercycler® gradient in un dispositivo in situ. Fornito con calotta trasparente.
Specifiche
  • Descrizione:
    Mastercycler® ep cycleManager, incl. installation instructions,
    comprehensive online-help, connection cable
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Adattatore in situ
    Adattatore in situ
    460-1325
    Log in per disponibilità
    1 * 1 ST - SFr. 1’490.00