Rack per provette, epMotion®
24 (4×6), 12ר40** mm, Acciaio inossidabile
Catalog # EPPE5075792.125
Supplier: EPPENDORF
Jump to:
Product Details & Documents
Stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
*screw cap tubes, ** HPLC vials.
Specifications
- Per provette:12ר40** mm
- N. di fori:24 (4×6)
- Materiale:Acciaio inossidabile