RAT STANDARD SERUM REFERENCE, BATCH 4227 1 * 0,5 mL
Catalog # ALPDNOR-03
Anbieter: Alpha Diagnostics International
RAT STANDARD SERUM REFERENCE, BATCH 4227 1 * 0,5 mL
Catalog # ALPDNOR-03
Anbieter: Alpha Diagnostics International
Frequently Bought Together
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Anbieter: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Sealing film for manual application, Parafilm® M
Anbieter: HEATHROW
VWR®, Salviette per il viso
Anbieter: VWR Collection
VWR®, Guanti in Nitrile
Anbieter: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Anbieter: VWR Chemicals
Strisce reattive pH, MQuant®, Supelco®
Anbieter: Merck
Guanti in nitrile, Kimtech™ Purple Nitrile™
Anbieter: KIMBERLY CLARK
VWR®, Pipette sierologiche, Standard Line
Anbieter: VWR Collection
VWR® NITRILE LIGHT, Guanti in nitrile
Anbieter: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Anbieter: VWR Collection