Nitrogen receivers (schlenk Tubes)
10 ml, - NS, 14/23 NS
Codice catalogo 201-1161
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and standard ground side stopcock with tube connector.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifiche
- Dimensione cono:- NS
- Cono femmina:14/23 NS
- Capacità:10 ml