Palloni per polveri
250 ml, 29/32 NS
Codice catalogo 201-0626
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Cono femmina:29/32 NS
- Capacità:250 ml