SRK® Surfaces and Equipment Microbial Testing Devices
Product Details & Documents
Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.
Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula
Compliance: SRK® kits are designed compliant with ISO 18593:2018, ISO 18593:2004, ISO 17604:2003, and EU directive 2001/471/E.C
Media choice for enhanced preservation: Choose between an isotonic solution or a wide range of selective and enrichment broths: Half Fraiser, RVS, BPW 1%, TSB, UNS, and Letheen broth
Sterile sampling frames: the definition of a set sampling area is essential for microbial bioburden validations
Sampling frames will make your surface sampling match that standard
This product line groups a variety of product codes, each optimised to address specific needs.
Documents
Specifiche
- Descrizione:SRK® - small foam sponge attached to a screw cap tube 16×100, without medium, in bulk; 50×6, bulk; 16×100 skirted sferic bottom tube
- Seq:24
- Colore tappo:blu