TIM-1 (C-TERM/INTERNAL) 1 * 0,1 mL
ALPDTIM11/12-S
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Antigen name:Protocadherin beta 16
- Cat. No.:BIRBORB3755-100
- Reactivity:Human
- Immunogen:Synthetic peptide
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Antigen symbol:PCDHB16
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



