TIM-1 (C-TERM/INTERNAL) 1 * 0,1 mg
Catalog # ALPDTIM11/12-A
Supplier: Alpha Diagnostics International
TIM-1 (C-TERM/INTERNAL) 1 * 0,1 mg
Catalog # ALPDTIM11/12-A
Supplier: Alpha Diagnostics International
TIM-1 (C-TERM/INTERNAL) 1 * 0,1 mg
Product Details & Documents
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
#: 20864.320
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
#: 83640.320
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., grado gradiente for liquid chromatography, Supelco®
#: 1.00030.2500
Supplier: Merck
Sodio idrossido 0.1 mol/l (0.1 N) in soluzione acquosa, AVS TITRINORM Reag. Ph. Eur. soluzione volumetrica
#: 31770.294
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
#: 1.09439.1000
Supplier: Merck



