Automatic burettes, Schilling pattern, Class B
25 ml, 0,1 ml
Codice catalogo KART1980
Nome del fornitore: KARTELL
Vai a
Product Details & Documents
PMP/PE, class B.
- Push button for drop-by- drop delivery
- PE reservoir with stable LDPE base
According to DIN 12.700 standards, class B
Certifications: According to DIN 12.700 standards, class B
Specifiche
- Capacità:25 ml
- Divisione:0,1 ml