Citidina
Glass bottle for solids, 10 g
Codice catalogo ROTH0717.1
Marketsource
Nome del fornitore: Roth Carl
Vai a
Product Details & Documents
Sinonimi: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
- Numero CAS:65-46-3
- Formula:C₉H₁₃N₃O₅
- Numero MDL:MFCD00006545
- Molecular Weight:243.22 g/mol
- temperatura di conservazione:Ambiente
- EINECS:200-610-9
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:10 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



