Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
EDTA

EDTA

25 kg
Catalog # ROTH8040.5
Irritant
Marketsource
Supplier:   Roth Carl
EDTA
Catalog # ROTH8040.5
Supplier:  Roth Carl
Marketsource
CAS Number:  60-00-4
EDTA
CAS Number:  60-00-4
Price
SFr. 2’148.00
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  • CAS Number:60-00-4
  • Formula:C₁₀H₁₆N₂O₈
  • MDL Number:MFCD00003541
  • Molecular Weight:292.25 g/mol
  • Merck Index:13,03546
  • Boiling Pt:614 °C (1013 hPa)
  • Melting Pt:220 °C
  • Density:0,86 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambiente
  • EINECS:200-449-4
  • REACH:01-2119486399-18
Specifications

Specifications

  • Conf:
    25 kg

Specifications