Orders LinkContactUsLinkComponent
Iodio
Iodio
Bottiglia di vetro, 250 g
Catalog # ROTHX864.2
Health
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Iodio
Catalog # ROTHX864.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7553-56-2
Iodio
Bottiglia di vetro, 250 g
CAS Number:  7553-56-2
Price
190.80 SFr.
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:13,05036
  • Boiling Pt:184,3 °C (1013 hPa)
  • Melting Pt:114 °C
  • Density:4,9 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:231-442-4
  • UN:3495
  • ADR:8,III
Specifications
Specifications
  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    250 g