Buckets, TruCLEAN™
TruCLEAN™ bucket, blu, 25 L
Codice catalogo 129-0105
Nome del fornitore: PERFEX
Vai a
Product Details & Documents
Temperature resistant polypropylene buckets with stainless steel handles.
- Buckets interlock when placed side by side
- Non-porous, super smooth surface prevents microbial growth
- Embossed graduations in English for accurate measure
- Stainless steel handles will not rust, corrode or oxidize
- Autoclavable, gamma ETO sterilization compatible
- Impervious to most chemicals, solvents and detergents
Buckets are seated firmly within cart base eliminating any possibility of spillage during transport.
Ideal for any critical manufacturing environment. Compatible with TruCLEAN Pro Mopping Systems.
Specifiche
- Capacità:25 L
- Colore:blu
- Descrizione:TruCLEAN™ bucket
- Seq:07