Magnetic stirring bars, PTFE, triangular, BOLA
130 mm, 38 mm, Recipienti di grandi dimensioni
Codice catalogo BOHLC357-30
Nome del fornitore: Bohlender
Magnetic stirring bars, PTFE, triangular, BOLA
Codice catalogo BOHLC357-30
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
Triangular shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE.
- Chemical resistant
Temperature resistance: −200 to +250 °C.
Most commonly used for big vessels in which strong turbulence can be acheived with low speeds. These can also been useful for mixing of reagents which resist dissolving and for avoiding any residues at the bottom of the vessels.
Specifiche
- Ø:38 mm
- Per:Recipienti di grandi dimensioni
- Lunghezza:130 mm
- Seq:10



