Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
High purity qualitative filter paper

High purity qualitative filter paper

1F, 104×250 mm
Codice catalogo 512-6182
Nome del fornitore:   Ahlstrom
High purity qualitative filter paper
Codice catalogo 512-6182
Nome del fornitore:  Ahlstrom
High purity qualitative filter paper
High purity qualitative filter paper

1F, 104×250 mm

Price
SFr. 64.70
Log in per disponibilità
1 * 100 ST
Product Details & Documents

Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Wet-strengthened grades contain a reinforcement agent that increases their resistance to rupture when wet, such as in vacuum filtration, and their resistance when filtering acidic solutions.

Grade 5 and V5: Very fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.
Grade 3: Fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.

Grade V150: Medium fast filtering, open to medium fiber matrix. For qualitative tasks, coarser precipitates such as lead-, iron- and silver sulfide; alkali carbonates; calcium oxalate.
Grade 1F and V1F: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulphate.

Grade Munktell no. 1 and 292: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate.
Grade Munktell no. 2 and 292a: Slowly filtering, tight fiber matrix. For filtration of fine precipitates such as lead dioxide, calcium fluoride, nickel sulfide, zinc sulfide.

Grade 106: Slowly filtering for fine precipitates such as lead dioxide, calcium fluoride, nickel sulphide and zinc sulphide.
Grade 120H and V120H: Very slowly filtering, tight fiber matrix. For fine precipitates such as barium sulphate, metastannic acid and cuprous oxide.

Specifiche
  • Grado:
    1F
  • Seq:
    23
  • Taglia:
    104×250 mm
  • Cat. No.:
    512-6182
  • Peso:
    80 g/m²
  • Contenuto di cenere:
    0,06%
  • Ritenzione delle particelle:
    5 - 6 µm
  • Velocità di filtrazione:
    300 s (Herzberg)