Heating tapes, IT-H
Heating tape IT-H with 0,5 m, 180 W output
Catalog # ISOP740644-000
Supplier: ISOPAD
Jump to:
Product Details & Documents
Isopad IT-H heating tape is suitable where high power input is needed or where work has to be carried out at high temperatures.
- Working element temperature of up to 900 °C
- Non-hazardous, ordinary area
- Storage temperature –40 to +50 °C
This tape should only be used in dry atmospheres with additional electrical protection and always with a temperature regulator.
Specifications
- Descrizione:Heating tape IT-H with 0,5 m, 180 W output
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



