Standard ground joints
- NS, 29/32 NS, 33 mm
Codice catalogo QUTW1033
Nome del fornitore: QUARTZTECHNISCHE WERKSTATTEN
Vai a
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.
Specifiche
- Dimensione cono:- NS
- Cono femmina:29/32 NS
- Ø esterno tubo:33 mm
- Seq:04