Standard ground joints
19/26 NS, - NS, 17 mm
Codice catalogo QUTW1045
Nome del fornitore: QUARTZTECHNISCHE WERKSTATTEN
Vai a
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.
Specifiche
- Dimensione cono:19/26 NS
- Cono femmina:- NS
- Ø esterno tubo:17 mm
- Seq:07