Orders LinkContactUsLinkComponent
Standard ground joints
Standard ground joints
29/32 NS, - NS, 26 mm
Codice catalogo QUTW1047
Nome del fornitore:   QUARTZTECHNISCHE WERKSTATTEN
Standard ground joints
Codice catalogo QUTW1047
Nome del fornitore:  QUARTZTECHNISCHE WERKSTATTEN
Standard ground joints
Standard ground joints
29/32 NS, - NS, 26 mm
Price
SFr. 199.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving high resistance to temperature changes.
Specifiche
  • Dimensione cono:
    29/32 NS
  • Cono femmina:
    - NS
  • Ø esterno tubo:
    26 mm
  • Seq:
    09