Heating tapes, IT-S20
Heating tape IT-S20, 2 m length, 440 W
Codice catalogo ISOP153620-000
Nome del fornitore: ISOPAD
Vai a
Product Details & Documents
Isopad IT-S20 is a glass silk, preterminated, flexible heating tape with a copper-tinned outer braiding serving as a protective earth braid. The temperature depends on the power and application. This enables maximum power to be installed even in the small surfaces. Provided with class I electrical protection.
- Maximum operating temperature: 450 °C
- Width×Thickness: 12×7 mm (±10%)
- Maximum power per meter 220 W/m
It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.
Specifiche
- Descrizione:Heating tape IT-S20, 2 m length, 440 W
- Seq:03



