Movable spine with pelvis
Movable spine with pelvis
Codice catalogo 763-0082
Marketsource
Nome del fornitore: 3B Scientific
Vai a
Product Details & Documents
Mounted on flexible stand pillar. With distal disc herniation between the 3rd and 4th vertebrae. Visible spinal nerves, blood vessels and discs. For true-to-life bends. Tripod included.
Specifiche
- Descrizione:Movable spine with pelvis