Labeling tapes, TX
Label printer tape, Poliestere, Thermal transfer, nero/giallo, PT-7000/8000, 18 mm×15 m
Catalog # BROTTX-641
Supplier: BROTHER
Jump to:
Product Details & Documents
Specifications
- Stampante:PT-7000/8000
- Printing method:Thermal transfer
- Colore:nero/giallo
- Descrizione:Label printer tape
- Dimensioni:18 mm×15 m
- Materiale:Poliestere
- Seq:16
- Cat. No.:BROTTX-641