Watch glasses
Watch glass, glass, with fused rim, 200 mm
Codice catalogo HECH40010020
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
- Good chemical restistance
- Available with or without hole
Certifications: DIN 12 341
Specifiche
- Ø:200 mm
- Descrizione:Watch glass, glass, with fused rim
- Seq:22