Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Powder funnel, short stem

Powder funnel, short stem

50 mm, 18 mm, 20 mm
Catalog # 221-1163
Powder funnel, short stem
Catalog # 221-1163
Powder funnel, short stem
Powder funnel, short stem

50 mm, 18 mm, 20 mm

Price
SFr. 10.10
Log in for availability
1 * 1 ST
Product Details & Documents

Soda-Kalkglas.

Specifications
  • Ø esterno imbuto:
    50 mm
  • Seq:
    02
  • Ø esterno stelo:
    18 mm
  • Lunghezza asta:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.