Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Glicina

Glicina

Bottiglia di vetro, 250 g
Catalog # ROTH3790.1
Marketsource
Supplier:   Roth Carl
Glicina
Catalog # ROTH3790.1
Supplier:  Roth Carl
Marketsource
CAS Number:  56-40-6
Glicina
Glicina

Bottiglia di vetro, 250 g

CAS Number:  56-40-6
Price
SFr. 24.60
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Boiling Pt:181 °C (1013 hPa)
  • Melting Pt:83 °C
  • Density:1,64 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:200-272-2
Specifications

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    250 g
  • Cat. No.:
    ROTH3790.1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.