Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Ion selective electrodes, bromide, Orion™

Ion selective electrodes, bromide, Orion™

Combinazione bromuro ionplus® Sure-Flow®, 9635BNWP, 1,0 - 5x10-6 M, 0...+80 °C, 9635BNWP, BNC impermeabile
Catalog # 662-1571
Supplier:   Thermo Orion
Ion selective electrodes, bromide, Orion™
Catalog # 662-1571
Supplier:  Thermo Orion
Ion selective electrodes, bromide, Orion™
Ion selective electrodes, bromide, Orion™

Combinazione bromuro ionplus® Sure-Flow®, 9635BNWP, 1,0 - 5x10-6 M, 0...+80 °C, 9635BNWP, BNC impermeabile

Price
SFr. 1’010.00
Log in for availability
1 * 1 ST
Product Details & Documents

These electrodes are designed for measuring bromide ions in aqueous solutions quickly, simply and economically.

  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode (9635BNWP) does not require a separate reference electrode, making it convenient to use with small sample volumes

Informations de commande: The half cell electrodes require a separate reference electrode which can be ordered separately. Screw cap electrode is supplied without cable.

Specifications
  • Concentrazione:
    1,0 - 5x10-6 M
  • Connettore:
    BNC impermeabile
  • Tipo:
    9635BNWP
  • Descrizione:
    Combinazione bromuro ionplus® Sure-Flow®, 9635BNWP
  • Seq:
    03
  • Temperatura:
    0...+80 °C
Accessories

Product Information

  • Catalog #
    Descrizione
    Concentrazione
    Conf
    Availability
    Price
  • ORIO943506
    Bromuro, soluzione standard
    0,1M
    475 mL
    Log in for availability
    1 * 475 mL - SFr. 136.00

Product Information

  • Catalog #
    Descrizione
    Tipo
    Tipo di conf.
    Availability
    Price
  • 662-0824
    Regolatore di forza ionica per elettrodo ione-selettivo per cloruro, in flacone
    Ion selective electrode reagents
    Bottle
    Log in for availability
    1 * 475 mL - SFr. 126.00

Product Information

  • Catalog #
    Descrizione
    Conf
    Availability
    Price
  • 662-1524
    Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    5
    Log in for availability
    1 * 5 ST - SFr. 150.00

Product Information

  • Catalog #
    Descrizione
    Conf
    Availability
    Price
  • 662-0750
    Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    5
    Log in for availability
    1 * 5 ST - SFr. 150.00

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.