Powder funnels, PP/PE
350 mm, 65 mm
Codice catalogo BURK9606-0350
Nome del fornitore: BURKLE
Vai a
Product Details & Documents
These powder funnels are designed for safe filling of containers.
- PP/PE
9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
Specifiche
- Ø esterno imbuto:350 mm
- Seq:04
- Ø esterno stelo:65 mm



