Pipette tips, PreCision Line
Standard, 1000 µl, -, 1000 µl pipettes, 50 bags of 1000 tips
Catalog # BZYM761013
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
: DIN 12650
Specifications
- Confezione:50 bags of 1000 tips
- Sterili:-
- Volume:1000 µl
- Seq:20
- Stile punta:Standard
- Per uso con:1000 µl pipettes