Pipette tips, PreCision Line
Standard, 1000 µl, -, 1000 µl pipettes, 10 bags of 1000 tips
Codice catalogo BZYM761011
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifiche
- Confezione:10 bags of 1000 tips
- Sterili:-
- Volume:1000 µl
- Seq:19
- Stile punta:Standard
- Per uso con:1000 µl pipettes