Pipette tips, PreCision Line
Standard, 200 µl, +, 200 µl pipettes, 10 racks of 96 tips
Codice catalogo BZYM760218
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifiche
- Confezione:10 racks of 96 tips
- Sterili:+
- Volume:200 µl
- Seq:15
- Stile punta:Standard
- Per uso con:200 µl pipettes