Pipette tips, PreCision Line
Standard, 200 µl, -, 200 µl pipettes, 50 bags of 1000 tips
Catalog # BZYM760213
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifications
- Confezione:50 bags of 1000 tips
- Sterili:-
- Volume:200 µl
- Seq:13
- Stile punta:Standard
- Per uso con:200 µl pipettes