Pipette tips, PreCision Line
Standard, 10 µl, -, 10 µl Sartorius Proline, Gilson P-2/P-10, Rainin Pipet-Classic PR2/PR10 + Pipet-Lite SL-2/SL-10, 50 bags of 1000 tips
Codice catalogo BZYM760013
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifiche
- Confezione:50 bags of 1000 tips
- Sterili:-
- Volume:10 µl
- Seq:06
- Stile punta:Standard
- Per uso con:10 µl Sartorius Proline, Gilson P-2/P-10, Rainin Pipet-Classic PR2/PR10 + Pipet-Lite SL-2/SL-10