Pipette tips, PreCision Line
Crystal, 10 µl, +, 10 µl Eppendorf, Brand and Socorex pipettes, 10 racks of 96 tips
Catalog # BZYM760008
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifications
- Confezione:10 racks of 96 tips
- Sterili:+
- Volume:10 µl
- Seq:04
- Stile punta:Crystal
- Per uso con:10 µl Eppendorf, Brand and Socorex pipettes