Pipette tips, PreCision Line
Crystal, 10 µl, -, 10 µl Eppendorf, Brand and Socorex pipettes, 50 bags of 1000 tips
Codice catalogo BZYM760003
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Specifiche
- Confezione:50 bags of 1000 tips
- Sterili:-
- Volume:10 µl
- Seq:02
- Stile punta:Crystal
- Per uso con:10 µl Eppendorf, Brand and Socorex pipettes