Vacuum Controller, V-850/V-855
Product Details & Documents
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option of reducing their water consumption by adding a cooling water valve.
V-855 vacuum controller:
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
Delivery information: Delivered with an air vent valve and pressure sensors.
Specifiche
- Descrizione:Controller del vuoto V-850
- Seq:01
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Cavo di comunicazione per R-210/R-215
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Woulff bottle for V-850/V-855
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Automatic distillation probe for V-855
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Assembly set for R-210/R-215
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Controllo remoto per V-850/V-855