VWR®, Rack per provette
27 (3×9), 8 mm, 180×60×45 mm, 45 mm
525-0192
PTFE.
- Eccellente resistenza chimica
- Resistenza termica da −200 a +280 °C
- Superfici antiadesione per una pulizia semplice
Robust and inert to virtually all chemical reagents, with very good thermal resistance. Suitable for use in cryogenics, freezers and ovens.
Documents
VWR® Lab bric-a-brac
Product Literature
- per Ø provette:8 mm
- N. di fori:27 (3×9)
- Altezza:45 mm
- L×P×A:180×60×45 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



