Blood dilution vials
25,00 ml, 35×56 mm, Yes, Bulk pack
Codice catalogo SIMPV130-1
Nome del fornitore: Simport Scientific
Vai a
Product Details & Documents
The dilution vials are made of PS with optically clear parallel sides. The snap cap is spill proof and can be easily removed.
Specifiche
- Tappo incluso:Yes
- Ø×A:35×56 mm
- Confezione:Bulk pack
- Capacità:25,00 ml
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



