Dropping bottles, Lucas
5 ml
Codice catalogo MUEL14.196.01
Nome del fornitore: MUELLER KREMPEL
Vai a
Product Details & Documents
Amber glass, without closure.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Please order screw caps and drop dispensers separately
Brown coloured to protect light-sensitive materials.
Specifiche
- Capacità:5 ml
- Seq:01