Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sample loops for Cheminert® valves, PEEK

Sample loops for Cheminert® valves, PEEK

Sample loop, PEEK, 250 µl, max. 240 bar
Codice catalogo JOURJR-SLC-85-250
Nome del fornitore:   VICI JOUR RESEARCH
Sample loops for Cheminert® valves, PEEK
Codice catalogo JOURJR-SLC-85-250
Nome del fornitore:  VICI JOUR RESEARCH
Sample loops for Cheminert® valves, PEEK

Sample loop, PEEK, 250 µl, max. 240 bar

Price
SFr. 92.40
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.

  • Fits all Cheminert® C1, C2, C2V, and C3 type valves
  • Tolerance of ±0,025 mm

PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.

Lieferinformationen: Supplied with PEEK hex-head nuts and PEEK ferrules.

Specifiche
  • Capacità:
    250 µl
  • Descrizione:
    Sample loop
  • Materiale:
    PEEK
  • Seq:
    07
  • Pressione:
    max. 240 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.