Sample loops for Cheminert® valves, PEEK
Sample loop, PEEK, 50 µl, max. 400 bar
Catalog # JOURJR-SLC-85-50
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.
- Fits all Cheminert® C1, C2, C2V, and C3 type valves
- Tolerance of ±0,025 mm
PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.
: Supplied with PEEK hex-head nuts and PEEK ferrules.
Specifications
- Capacità:50 µl
- Descrizione:Sample loop
- Materiale:PEEK
- Seq:04
- Pressione:max. 400 bar



