Unions
Union, through bore LP body only, 1/4”-28, for flanged tubing, Poliammide
Codice catalogo JOURJR-060-5
Nome del fornitore: VICI JOUR RESEARCH
Vai a
Product Details & Documents
Unions for easy connection of low pressure solvent lines.
- Low dead volume
- Inert and biocompatible
Specifiche
- Descrizione:Union, through bore LP body only, 1/4”-28, for flanged tubing
- Materiale:Poliammide
- Seq:01