Orders LinkContactUsLinkComponent
Unions
Unions
Union, LP 0,75 mm bore, complete, 1/16” tubing, PEEK
Catalog # JOURJR-1068
Supplier:   VICI JOUR RESEARCH
Unions
Catalog # JOURJR-1068
Supplier:  VICI JOUR RESEARCH
Unions
Union, LP 0,75 mm bore, complete, 1/16” tubing, PEEK
Price
SFr. 60.20
Log in for availability
1 * 1 ST
Product Details & Documents
Unions for easy connection of low pressure solvent lines.
  • Low dead volume
  • Inert and biocompatible
Specifications
  • Descrizione:
    Union, LP 0,75 mm bore, complete, 1/16” tubing
  • Materiale:
    PEEK
  • Seq:
    06