Unions
Union, LP 0,75 mm bore, complete, 1/16” tubing, PEEK
Catalog # JOURJR-1068
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
Unions for easy connection of low pressure solvent lines.
- Low dead volume
- Inert and biocompatible
Specifications
- Descrizione:Union, LP 0,75 mm bore, complete, 1/16” tubing
- Materiale:PEEK
- Seq:06