Heating tapes, HBRC
Self-limiting heating tape with low working temperature, width 11 mm, 26 W
Codice catalogo HORS021403
Nome del fornitore: HORST
Vai a
Product Details & Documents
Selflimiting heating tape is suitable for plastic pipes and plastic containers. It comes with an additional protective braid and humidity proof.
- Nominal temperature 65 °C
- With min. bending radius of 20 mm
Thickness: 5,5 mm
Specifiche
- Descrizione:Self-limiting heating tape with low working temperature, width 11 mm, 26 W
- Seq:01