Weighing boats, rounded
3,0 ml, 85 mm
Codice catalogo NEUB1261-17210
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Weighing boats, rounded
Codice catalogo NEUB1261-17210
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Vai a
Product Details & Documents
Weighing funnels, borosilicate glass 3.3, designed to weigh chemicals or for weighing chemicals with tubular stalk.
- Excellent chemical resistance
- Excellent temperature Resistance
Specifiche
- Capacità:3,0 ml
- Lunghezza:85 mm
- Seq:03