Distilling bridges
200 mm, 19/26 NS
Codice catalogo WITG1.205.260
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Specifiche
- Dimensione cono:19/26 NS
- Lunghezza mantello:200 mm