Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
150 mm, 14/23 NS
Codice catalogo WITG1.205.150
Nome del fornitore:   witeg Labortechnik
Distilling bridges
Codice catalogo WITG1.205.150
Nome del fornitore:  witeg Labortechnik
Distilling bridges
Distilling bridges
150 mm, 14/23 NS
Price
174.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Specifiche
  • Dimensione cono:
    14/23 NS
  • Lunghezza mantello:
    150 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.