Orders LinkContactUsLinkComponent
Lysates, human
Lysates, human
HEK293, whole cell lysate, 500 µG
Codice catalogo ROCKW09-000-365
Nome del fornitore:   Rockland Immunochemicals
Lysates, human
Codice catalogo ROCKW09-000-365
Nome del fornitore:  Rockland Immunochemicals
Numero CAS:  
Lysates, human
Lysates, human
HEK293, whole cell lysate, 500 µG
Numero CAS:  
Price
SFr. 259.00
Log in per disponibilità
1 * 500 µG
Product Details & Documents
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
  • Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.
Specifiche
Specifiche
  • Conf:
    500 µG
  • Descrizione:
    HEK293, whole cell lysate